rel-(3aS,6aS)-Hexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide hydrochloride 97%

Supplier: Ambeed
Warning

A1492704-100MG A1492704-250MG
76665-918EA 205 USD
76665-918 76665-920
rel-(3aS,6aS)-Hexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide hydrochloride 97%
rel-(3aS,6aS)-Hexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂S
CAS Number: 1320326-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR