rel-(3aS,6aS)-5-Benzylhexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide 97%

Supplier: Ambeed
Warning

A1492701-100MG A1492701-250MG
76665-914EA 175 USD
76665-914 76665-916
rel-(3aS,6aS)-5-Benzylhexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide 97%
rel-(3aS,6aS)-5-Benzylhexahydro-2H-thieno[2,3-c]pyrrole 1,1-dioxide

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂S
CAS Number: 1320326-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR