rel-Methyl (1R,5R)-3-benzyl-3-azabicyclo[3.2.0]heptane-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A1492629-100MG
A1492629-250MG
76665-874EA
40
USD
76665-874
76665-876
rel-Methyl (1R,5R)-3-benzyl-3-azabicyclo[3.2.0]heptane-1-carboxylate 97%
rel-Methyl (1R,5R)-3-benzyl-3-azabicyclo[3.2.0]heptane-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₂ |
CAS Number:
2751265-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...