1-Cyclooctyl-3-mesityl-1H-imidazol-3-ium chloride 97%
Supplier: Ambeed
|
Warning
|
A1492624-100MG
A1492624-1G
A1492624-250MG
76665-868EA
41.38
USD
76665-868
76665-870
76665-872
1-Cyclooctyl-3-mesityl-1H-imidazol-3-ium chloride 97%
1-Cyclooctyl-3-mesityl-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₂₀H₂₉ClN₂ MW: 332.9107 g/mol |
CAS Number:
1583244-05-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...