1-Cycloheptyl-3-mesityl-1H-imidazol-3-ium chloride 97%
Supplier: Ambeed
|
Warning
|
A1492623-100MG
A1492623-1G
A1492623-250MG
76665-862EA
59.42
USD
76665-862
76665-864
76665-866
1-Cycloheptyl-3-mesityl-1H-imidazol-3-ium chloride 97%
1-Cycloheptyl-3-mesityl-1H-imidazol-3-ium chloride
Precaución: for Research Use Only
Formula:
C₁₉H₂₇ClN₂ MW: 318.8841 g/mol |
CAS Number:
1583244-04-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...