1-Cyclopentyl-3-mesityl-1H-imidazol-3-ium chloride 97%

Supplier: Ambeed
Warning

A1492621-100MG A1492621-1G A1492621-250MG
76665-850EA 54.11 USD
76665-850 76665-852 76665-854
1-Cyclopentyl-3-mesityl-1H-imidazol-3-ium chloride 97%
1-Cyclopentyl-3-mesityl-1H-imidazol-3-ium chloride

Precaución: for Research Use Only

Formula: C₁₇H₂₃ClN₂
MW: 290.8309 g/mol
CAS Number: 1583244-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR