(S)-3,3-Dimethyl-2-(2,2,2-trifluoroacetamido)butanoic acid 98%

Supplier: Ambeed
Warning

A1491995-1G A1491995-250MG A1491995-5G A1491995-100MG
76665-842EA 8 USD
76665-842 76665-844 76665-846 77875-474
(S)-3,3-Dimethyl-2-(2,2,2-trifluoroacetamido)butanoic acid 98%
(S)-3,3-Dimethyl-2-(2,2,2-trifluoroacetamido)butanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₂F₃NO₃
MDL Number: MFCD12795957
CAS Number: 666832-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR