1-Butyl-3-vinyl-1H-imidazol-3-ium bromide 98%

Supplier: Ambeed
Warning

A1490884-1G A1490884-25G A1490884-5G
76665-836EA 5.31 USD
76665-836 76665-838 76665-840
1-Butyl-3-vinyl-1H-imidazol-3-ium bromide 98%
1-Butyl-3-vinyl-1H-imidazol-3-ium bromide

Precaución: for Research Use Only

Formula: C₉H₁₅BrN₂
MW: 231.13 g/mol
MDL Number: MFCD18251314
CAS Number: 34311-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR