3-Chloro-2,4(3H,5H)-furandione 99%

Supplier: Ambeed
Warning

Synonyms: 3-Chlorotetronic Acid, 3-Chlorofuran-2,4(3H,5H)-dione, 3-Chlorotetrahydrofuran-2,4-dione, 3-Chloro-4-hydroxy-2(5H)-furanone

A1490694-100G A1490694-1G A1490694-25G A1490694-5G
76665-808EA 595.17 USD
76665-808 76665-810 76665-812 76665-814
3-Chloro-2,4(3H,5H)-furandione 99%
3-Chloro-2,4(3H,5H)-furandione

Precaución: for Research Use Only

Formula: C₄H₃ClO₃
MW: 134.52 g/mol
MDL Number: MFCD00134273
CAS Number: 4971-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR