4,4'-Dimethyl-trans-stilbene 99+%

Supplier: Ambeed
Warning

Synonyms: (E)-1,2-Di-p-tolylethene

A1490692-1G A1490692-5G
76665-804EA 93.36 USD
76665-804 76665-806
4,4'-Dimethyl-trans-stilbene 99+%
4,4'-Dimethyl-trans-stilbene

Precaución: for Research Use Only

Formula: C₁₆H₁₆
MW: 208.2982 g/mol
MDL Number: MFCD00136904
CAS Number: 18869-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR