(Ir[Me(Me)ppy]2(5,5'-dCF3bpy))PF6 97%

Supplier: Ambeed
Warning

A1490574-100MG A1490574-250MG A1490574-25MG A1490574-50MG
76665-780EA 88 USD
76665-780 76665-782 76665-784 76665-786
(Ir[Me(Me)ppy]2(5,5'-dCF3bpy))PF6 97%
(Ir[Me(Me)ppy]2(5,5'-dCF3bpy))PF6

Precaución: for Research Use Only

Formula: C₃₈H₃₀F₁₂IrN₄P
CAS Number: 2828433-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR