4-Bromo-3-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A1490441-1G A1490441-25G A1490441-5G
76665-760EA 27.59 USD
76665-760 76665-762 77567-108
4-Bromo-3-hydroxybenzonitrile 98%
4-Bromo-3-hydroxybenzonitrile

Precaución: For research use only.

Formula: C₇H₄BrNO
MW: 198.017 g/mol
MDL Number: MFCD16999194
CAS Number: 916213-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR