16-Bromopalmitic acid 98%

Supplier: Ambeed
Warning

A1490308-100G A1490308-1G A1490308-5G A1490308-25G
76665-744EA 2475 USD
76665-744 76665-746 76665-748 77624-492
16-Bromopalmitic acid 98%
16-Bromopalmitic acid

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₁₄COOH
MDL Number: MFCD00040779
CAS Number: 2536-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR