18-Methoxy-18-oxooctadecanoic acid 98%

Supplier: Ambeed
Warning

A1490302-1G A1490302-250MG A1490302-25G A1490302-5G
76665-732EA 80 USD
76665-732 76665-734 76665-736 76665-738
18-Methoxy-18-oxooctadecanoic acid 98%
18-Methoxy-18-oxooctadecanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₆O₄
MDL Number: MFCD00673469
CAS Number: 72849-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR