19-(tert-Butoxy)-19-oxononadecanoic acid 98+%

Supplier: Ambeed
Warning

A1490290-1G A1490290-250MG A1490290-5G
76665-718EA 26 USD
76665-718 76665-720 77624-484
19-(tert-Butoxy)-19-oxononadecanoic acid 98+%
19-(tert-Butoxy)-19-oxononadecanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₄₄O₄
MDL Number: MFCD30737821
CAS Number: 1643852-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR