18-(tert-Butoxy)-18-oxooctadecanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl Hydrogen Octadecanedioate, Mono-tert-butyl Octadecanedioate, tBu ester-C16-acid, Octadecanedioic acid Mono-tert-butyl ester

A1490288-100G A1490288-1G A1490288-25G A1490288-5G
76665-710EA 647 USD
76665-710 76665-712 76665-714 76665-716
18-(tert-Butoxy)-18-oxooctadecanoic acid 98%
18-(tert-Butoxy)-18-oxooctadecanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₄
MDL Number: MFCD09991735
CAS Number: 843666-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR