tert-Butyl Hydrogen Hexadecanedioate 99%

Supplier: Ambeed
Warning

Synonyms: 16-(tert-Butoxy)-16-oxohexadecanoic acid, Hexadecanedioic acid Mono-tert-butyl ester, Boc-C14-COOH

A1490284-100G A1490284-1G A1490284-25G A1490284-500G A1490284-5G
76665-690EA 565 USD
76665-690 76665-692 76665-694 76665-696 76665-698
tert-Butyl Hydrogen Hexadecanedioate 99%
tert-Butyl Hydrogen Hexadecanedioate

Precaución: for Research Use Only

Formula: C₂₀H₃₈O₄
MDL Number: MFCD23136039
CAS Number: 843666-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR