11-(tert-Butoxy)-11-oxoundecanoic acid 98+%

Supplier: Ambeed
Warning

A1490276-100G A1490276-1G A1490276-25G A1490276-5G
76665-672EA 636.54 USD
76665-672 76665-674 76665-676 76665-678
11-(tert-Butoxy)-11-oxoundecanoic acid 98+%
11-(tert-Butoxy)-11-oxoundecanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₈O₄
MW: 272.38 g/mol
MDL Number: MFCD30737824
CAS Number: 1789702-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR