20-(tert-Butoxy)-20-oxoicosanoic acid 98%

Supplier: Ambeed
Warning

A1490258-100G A1490258-1G A1490258-25G A1490258-5G A1490258-500G
76665-640EA 730 USD
76665-640 76665-642 76665-644 76665-646 77875-438
20-(tert-Butoxy)-20-oxoicosanoic acid 98%
20-(tert-Butoxy)-20-oxoicosanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₄₆O₄
MDL Number: MFCD23136041
CAS Number: 683239-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR