1-Hydroxycyclohexyl phenyl ketone 99+%

Supplier: Ambeed
Warning

Synonyms: (1-Hydroxycyclohexyl)(phenyl)methanone

A1490211-1000G A1490211-100G A1490211-500G
76665-634EA 29 USD
76665-634 76665-636 76665-638
1-Hydroxycyclohexyl phenyl ketone 99+%
1-Hydroxycyclohexyl phenyl ketone

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MDL Number: MFCD00059561
CAS Number: 947-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR