2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 99+%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1-(4-(2-hydroxyethoxy)phenyl)-2-methylpropan-1-one

A1490209-100G A1490209-500G
76665-630EA 40.32 USD
76665-630 76665-632
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 99+%
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.2530 g/mol
MDL Number: MFCD00085267
CAS Number: 106797-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR