Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide 99%

Supplier: Ambeed
Danger

Synonyms: (Diphenylphosphoryl)(mesityl)methanone

A1490207-25G A1490207-100G A1490207-1000G
76665-628EA 8.49 USD
76665-628 77597-652 77624-482
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide 99%
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
MDL Number: MFCD00192110
CAS Number: 75980-60-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR