cis-Dichlorobis(pyridine)platinum(II) Pt 45.98%

Supplier: Ambeed
Danger

A1490035-100MG A1490035-1G A1490035-250MG
76665-622EA 45 USD
76665-622 76665-624 76665-626
cis-Dichlorobis(pyridine)platinum(II) Pt 45.98%
cis-Dichlorobis(pyridine)platinum(II)

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₂Pt
MW: 424.19 g/mol
MDL Number: MFCD00058858
CAS Number: 15227-42-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR