N,N-Dimethyl-[1,3'-biazetidin]-3-amine 2,2,2-trifluoroacetate 95+%

Supplier: Ambeed
Warning

A1489980-100MG A1489980-1G A1489980-250MG
76665-608EA 28.65 USD
76665-608 76665-610 76665-612
N,N-Dimethyl-[1,3'-biazetidin]-3-amine 2,2,2-trifluoroacetate 95+%
N,N-Dimethyl-[1,3'-biazetidin]-3-amine 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₀H₁₈F₃N₃O₂
MW: 269.2640 g/mol
CAS Number: 2757730-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR