4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-amine 97%
Supplier: Ambeed
|
Warning
|
A1489917-100MG
A1489917-1G
A1489917-250MG
76665-596EA
31.83
USD
76665-596
76665-598
76665-600
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-amine 97%
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-amine
Precaución: for Research Use Only
Formula:
C₁₈H₂₂BNO₂ MW: 295.1838 g/mol |
CAS Number:
1008788-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...