4',5'-Bis(4-aminophenyl)-[1,1':2',1''-terphenyl]-4,4''-diamine 97%
Supplier: Ambeed
|
Warning
|
A1489916-100MG
A1489916-250MG
A1489916-5G
A1489916-1G
76665-590EA
12
USD
76665-590
76665-592
76665-594
77597-650
4',5'-Bis(4-aminophenyl)-[1,1':2',1''-terphenyl]-4,4''-diamine 97%
4',5'-Bis(4-aminophenyl)-[1,1':2',1''-terphenyl]-4,4''-diamine
Precaución: for Research Use Only
Formula:
C₃₀H₂₆N₄ |
CAS Number:
2458125-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...