[1,1':3',1''-Terphenyl]-3,3'',5'-tricarboxylic acid 97%

Supplier: Ambeed
Warning

A1489914-100MG A1489914-1G A1489914-250MG
76665-578EA 21.22 USD
76665-578 76665-580 76665-582
[1,1':3',1''-Terphenyl]-3,3'',5'-tricarboxylic acid 97%
[1,1':3',1''-Terphenyl]-3,3'',5'-tricarboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₄O₆
MW: 362.33 g/mol
CAS Number: 2127158-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR