1,1,2,2-Tetrakis(4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl)ethene 97%

Supplier: Ambeed
Warning

A1489905-100MG A1489905-1G A1489905-250MG
76665-566EA 128.37 USD
76665-566 76665-568 76665-570
1,1,2,2-Tetrakis(4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl)ethene 97%
1,1,2,2-Tetrakis(4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl)ethene

Precaución: for Research Use Only

Formula: C₇₀H₄₈N₄
MW: 945.157 g/mol
MDL Number: MFCD34473821
CAS Number: 2424054-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR