5,5'-(1,4-Phenylenebis(ethyne-2,1-diyl))diisophthalic acid 97%

Supplier: Ambeed
Warning

A1489900-100MG A1489900-250MG
76665-554EA 80.63 USD
76665-554 76665-556
5,5'-(1,4-Phenylenebis(ethyne-2,1-diyl))diisophthalic acid 97%
5,5'-(1,4-Phenylenebis(ethyne-2,1-diyl))diisophthalic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₄O₈
MW: 454.3846 g/mol
CAS Number: 957014-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR