[1,1':4',1''-Terphenyl]-3,3'',5,5''-tetracarbaldehyde 97%

Supplier: Ambeed
Warning

A1489883-100MG A1489883-1G A1489883-250MG
76665-542EA 20 USD
76665-542 76665-544 76665-546
[1,1':4',1''-Terphenyl]-3,3'',5,5''-tetracarbaldehyde 97%
[1,1':4',1''-Terphenyl]-3,3'',5,5''-tetracarbaldehyde

Precaución: for Research Use Only

Formula: C₂₂H₁₄O₄
CAS Number: 2380275-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR