5,5'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)bis(thiophene-2-carbaldehyde) 97%

Supplier: Ambeed
Warning

A1489846-100MG A1489846-1G A1489846-250MG
76665-520EA 38.2 USD
76665-520 76665-522 76665-524
5,5'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)bis(thiophene-2-carbaldehyde) 97%
5,5'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)bis(thiophene-2-carbaldehyde)

Precaución: for Research Use Only

Formula: C₁₆H₈N₂O₂S₃
MW: 356.4419 g/mol
CAS Number: 882303-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR