5',5''''-(Anthracene-9,10-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid)) 97%

Supplier: Ambeed
Warning

A1489831-100MG A1489831-1G A1489831-250MG
76665-502EA 45.62 USD
76665-502 76665-504 76665-506
5',5''''-(Anthracene-9,10-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid)) 97%
5',5''''-(Anthracene-9,10-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid))

Precaución: for Research Use Only

Formula: C₅₄H₃₄O₈
MW: 810.8430 g/mol
CAS Number: 913343-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR