1,1,1-Trifluoro-N-((11bR)-4-oxido-2,6-bis(4-(trifluoromethyl)phenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A1489595-100MG A1489595-1G A1489595-250MG A1489595-50MG
76665-462EA 187.78 USD
76665-462 76665-464 76665-466 76665-468
1,1,1-Trifluoro-N-((11bR)-4-oxido-2,6-bis(4-(trifluoromethyl)phenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide 98%
1,1,1-Trifluoro-N-((11bR)-4-oxido-2,6-bis(4-(trifluoromethyl)phenyl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₃₅H₂₇F₉NO₅PS
MW: 775.61 g/mol
CAS Number: 2757287-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR