(11bR)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 98%

Supplier: Ambeed
Warning

A1489593-100MG A1489593-1G A1489593-250MG A1489593-50MG
76665-454EA 135.8 USD
76665-454 76665-456 76665-458 76665-460
(11bR)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide 98%
(11bR)-N-(2,6-Bis(4-trifluoromethylphenyl)-4-oxidodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-yl)-1,1,1-trifluoromethanesulfonamide

Precaución: for Research Use Only

Formula: C₃₅H₁₉F₉NO₅PS
MW: 767.5535 g/mol
CAS Number: 2757287-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR