1,4-Phenylenebis(methylene) didodecyl bis(carbonotrithioate) 98%

Supplier: Ambeed
Warning

A1489584-100MG A1489584-250MG A1489584-1G
76665-442EA 42.44 USD
76665-442 76665-444 77624-464
1,4-Phenylenebis(methylene) didodecyl bis(carbonotrithioate) 98%
1,4-Phenylenebis(methylene) didodecyl bis(carbonotrithioate)

Precaución: for Research Use Only

Formula: C₃₄H₅₈S₆
MW: 659.2143 g/mol
MDL Number: MFCD32174047
CAS Number: 960256-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR