[1,1'-Biphenyl]-4-yl(3-methoxyphenyl)methanone 98+%

Supplier: Ambeed
Warning

A1488799-25G A1488799-5G
76665-424EA 8.49 USD
76665-424 76665-426
[1,1'-Biphenyl]-4-yl(3-methoxyphenyl)methanone 98+%
[1,1'-Biphenyl]-4-yl(3-methoxyphenyl)methanone

Precaución: for Research Use Only

Formula: C₂₀H₁₆O₂
MW: 288.3399 g/mol
MDL Number: MFCD00812571
CAS Number: 79447-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR