[1,1'-Binaphthalene]-4,4'-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1488569-100MG A1488569-250MG
76665-418EA 49.87 USD
76665-418 76665-420
[1,1'-Binaphthalene]-4,4'-dicarboxylic acid 98%
[1,1'-Binaphthalene]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₄O₄
MW: 342.3442 g/mol
CAS Number: 123524-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR