(1S)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(dicyclohexylphosphane) 97%
Supplier: Ambeed
|
Warning
|
A1488512-100MG
A1488512-1G
A1488512-250MG
A1488512-50MG
76665-412EA
138
USD
76665-412
76665-414
76665-416
77875-408
(1S)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(dicyclohexylphosphane) 97%
(1S)-(4,4',6,6'-Tetramethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(dicyclohexylphosphane)
Precaución: for Research Use Only
Formula:
C₄₀H₆₀O₄P₂ |
CAS Number:
2829282-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...