5-Bromo-N-methylpicolinamide 95%

Supplier: Ambeed
Warning

A1488333-100MG A1488333-1G A1488333-250MG A1488333-5G
76665-404EA 9 USD
76665-404 76665-406 76665-408 76665-410
5-Bromo-N-methylpicolinamide 95%
5-Bromo-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MDL Number: MFCD07357421
CAS Number: 845305-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR