7-Nitro-1-tetralone 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Dihydro-7-nitronaphthalen-1(2H)-one, 7-Nitro-3,4-dihydronaphthalen-1(2H)-one
A1488255-100G
A1488255-1G
A1488255-25G
A1488255-5G
76665-390EA
785.07
USD
76665-390
76665-392
76665-394
76665-396
7-Nitro-1-tetralone 98+%
7-Nitro-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol |
MDL Number:
MFCD00019661 CAS Number: 40353-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...