2-(Dimethylamino)pyridine 98+%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethylpyridin-2-amine

A1488249-100G A1488249-25G A1488249-5G
76665-384EA 192.03 USD
76665-384 76665-386 76665-388
2-(Dimethylamino)pyridine 98+%
2-(Dimethylamino)pyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00006261
CAS Number: 5683-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR