Spiro[4.5]decan-7-one 98%

Supplier: Ambeed
Warning

A1488060-100MG A1488060-1G A1488060-250MG
76665-378EA 140.04 USD
76665-378 76665-380 76665-382
Spiro[4.5]decan-7-one 98%
Spiro[4.5]decan-7-one

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MW: 152.2334 g/mol
MDL Number: MFCD24688439
CAS Number: 62788-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR