3-Oxaspiro[5.5]undecan-8-one 98%

Supplier: Ambeed
Warning

A1488059-100MG A1488059-1G A1488059-250MG A1488059-50MG
76665-370EA 53 USD
76665-370 76665-372 76665-374 76665-376
3-Oxaspiro[5.5]undecan-8-one 98%
3-Oxaspiro[5.5]undecan-8-one

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₂
CAS Number: 2385721-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR