4-Methoxyphenylhydrazinium chloride 98%

Supplier: Ambeed
Warning

A1487830-25G A1487830-500G A1487830-5G A1487830-1G
76665-298EA 34 USD
76665-298 76665-300 76665-302 77875-396
4-Methoxyphenylhydrazinium chloride 98%
4-Methoxyphenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MDL Number: MFCD00012945
CAS Number: 19501-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR