8-Bromo-2-chloroquinazolin-4-amine 98+%

Supplier: Ambeed
Warning

A1487826-100G A1487826-1G A1487826-5G A1487826-25G
76665-292EA 1004 USD
76665-292 76665-294 76665-296 77597-614
8-Bromo-2-chloroquinazolin-4-amine 98+%
8-Bromo-2-chloroquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₈H₅BrClN₃
MDL Number: MFCD11858275
CAS Number: 956100-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR