[Ir(ppy)2(4-([2,2'-bipyridin]-4-yl)butanoic acid)][PF6] 98%

Supplier: Ambeed
Warning

A1487624-100MG A1487624-250MG A1487624-50MG
76665-266EA 241.89 USD
76665-266 76665-268 76665-270
[Ir(ppy)2(4-([2,2'-bipyridin]-4-yl)butanoic acid)][PF6] 98%
[Ir(ppy)2(4-([2,2'-bipyridin]-4-yl)butanoic acid)][PF6]

Precaución: for Research Use Only

Formula: C₃₆H₃₀F₆IrN₄O₂P
MW: 887.8302 g/mol
CAS Number: 2828447-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR