2-(2,6-Dioxopiperidin-3-yl)-5-fluoro-6-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride 95%

Supplier: Ambeed
Warning

A1487434-100MG A1487434-1G A1487434-250MG A1487434-5G
76665-248EA 91 USD
76665-248 76665-250 76665-252 76665-254
2-(2,6-Dioxopiperidin-3-yl)-5-fluoro-6-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride 95%
2-(2,6-Dioxopiperidin-3-yl)-5-fluoro-6-(piperazin-1-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₈ClFN₄O₄
MW: 396.80 g/mol
MDL Number: MFCD32203462
CAS Number: 2222114-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR