(±)-1,2-Dodecanediol 95%, (Chunks or flakes)

Supplier: Ambeed
Warning

A1487430-25G A1487430-5G
76665-244EA 44 USD
76665-244 76665-246
(±)-1,2-Dodecanediol 95%, (Chunks or flakes)
(±)-1,2-Dodecanediol

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₂
MDL Number: MFCD00004726
CAS Number: 1119-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR