4,7-Bis(5-bromothiophen-2-yl)-2λ4δ2-benzo[1,2-c;4,5-c']bis[1,2,5]thiadiazol 97%

Supplier: Ambeed
Warning

A1486684-100MG A1486684-250MG
76665-234EA 88.06 USD
76665-234 76665-236
4,7-Bis(5-bromothiophen-2-yl)-2λ4δ2-benzo[1,2-c;4,5-c']bis[1,2,5]thiadiazol 97%
4,7-Bis(5-bromothiophen-2-yl)-2λ4δ2-benzo[1,2-c;4,5-c']bis[1,2,5]thiadiazol

Precaución: for Research Use Only

Formula: C₁₄H₄Br₂N₄S₄
MW: 516.28 g/mol
CAS Number: 886746-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR