1,3,6,8-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 97%

Supplier: Ambeed
Warning

A1486677-100MG A1486677-1G A1486677-250MG
76665-228EA 42.44 USD
76665-228 76665-230 76665-232
1,3,6,8-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 97%
1,3,6,8-Tetrakis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene

Precaución: for Research Use Only

Formula: C₄₀H₅₄B₄O₈
MW: 706.0960 g/mol
MDL Number: MFCD34469727
CAS Number: 1398053-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR